Assay Details
Target Gene Details
Entrez Gene ID: | 12632 |
Gene Name: | cofilin 2, muscle |
Gene Aliases: |
- |
Location: |
Chr.12:54858815-54862877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cfl2 | NM_007688.2 | NP_031714.1 | ||
| AK004659.1 | ||||
| AK147174.1 | BAE27737.1 | |||
| BC007138.1 | AAH07138.1 | |||
| L29468.1 | AAA37433.1 |
More Information
Set Membership: |
|

Genomic Map