Assay Details
Target Gene Details
Entrez Gene ID: | 668158 |
Gene Name: | coiled-coil domain containing 85C |
Gene Aliases: |
EG668158, Gm265, Gm9010 |
Location: |
Chr.12:108203599-108275671 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc85c | NM_001159910.1 | NP_001153382.1 |

Genomic Map