Assay Details
Target Gene Details
Entrez Gene ID: | 77480 |
Gene Name: | kinase D-interacting substrate 220 |
Gene Aliases: |
3110039L19Rik, AI194387, AI316525, C330002I19Rik, mKIAA1250 |
Location: |
Chr.12:24974876-25063152 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kidins220 | NM_001081378.1 | 10 | 1110 | NP_001074847.1 |
| AK083260.1 | 4 | 434 | BAE43383.1 | |
| AK122478.1 | 7 | 700 | BAC65760.1 | |
| BC044772.2 | 5 | 466 | AAH44772.1 | |
| BC046467.1 | 5 | 466 | AAH46467.1 | |
| BC157898.1 | 10 | 994 |

Genomic Map