Assay Details
Target Gene Details
Entrez Gene ID: | 217588 |
Gene Name: | MAP3K12 binding inhibitory protein 1 |
Gene Aliases: |
4933408E06Rik |
Location: |
Chr.12:56328306-56345894 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mbip | NM_145442.2 | NP_663417.1 | ||
| AK077125.1 | BAC36628.1 | |||
| AK154318.1 | BAE32511.1 | |||
| AK171164.1 | BAE42286.1 | |||
| BC002277.1 | AAH02277.1 |
More Information
Set Membership: |
|

Genomic Map