Assay Details
Target Gene Details
Entrez Gene ID: | 12062 |
Gene Name: | bradykinin receptor, beta 2 |
Gene Aliases: |
B(2), B2, B2R, BK-2, BK2, BK2R, BKR2, BRB2 |
Location: |
Chr.12:105563172-105595244 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bdkrb2 | NM_009747.2 | NP_033877.3 | ||
| AK132469.1 | ||||
| BC137755.1 | ||||
| BC137756.1 | ||||
| X69682.1 | CAA49362.1 | |||
| X78438.1 | CAA55202.1 |
More Information
Set Membership: |
|

Genomic Map