Assay Details
Target Gene Details
Entrez Gene ID: | 13983 |
Gene Name: | estrogen receptor 2 (beta) |
Gene Aliases: |
ER[b], ERbeta, Estrb |
Location: |
Chr.12:76120419-76177259 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esr2 | NM_010157.3 | NP_034287.3 | ||
| NM_207707.1 | NP_997590.1 | |||
| NR_104386.1 | ||||
| AK054290.1 | BAC35719.1 | |||
| AK054301.1 | ||||
| AK054413.1 | BAC35770.1 | |||
| AY054413.1 | AAL15175.1 | |||
| BC141075.1 | ||||
| BC145329.1 | ||||
| U81451.1 | AAB51132.1 |

Genomic Map