Assay Details
Target Gene Details
Entrez Gene ID: | 77480 |
Gene Name: | kinase D-interacting substrate 220 |
Gene Aliases: |
3110039L19Rik, AI194387, AI316525, C330002I19Rik, mKIAA1250 |
Location: |
Chr.12:24974876-25063152 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kidins220 | NM_001081378.1 | NP_001074847.1 | ||
| AK021172.1 | ||||
| BC157898.1 |

Genomic Map