Assay Details
Target Gene Details
Entrez Gene ID: | 14281 |
Gene Name: | FBJ osteosarcoma oncogene |
Gene Aliases: |
D12Rfj1, c-fos, cFos |
Location: |
Chr.12:85473901-85477269 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fos | NM_010234.2 | NP_034364.1 | ||
| AK154418.1 | BAE32571.1 | |||
| BC029814.1 | AAH29814.1 |
More Information
Set Membership: |
|

Genomic Map