Assay Details
Target Gene Details
Entrez Gene ID: | 214305 |
Gene Name: | hedgehog interacting protein-like 1 |
Gene Aliases: |
1600002O04Rik, AK129450, mKIAA1822 |
Location: |
Chr.12:108305610-108328300 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hhipl1 | NM_001044380.1 | NP_001037845.1 | ||
| AK138646.1 | BAE23730.1 | |||
| BC113163.1 | ||||
| BC113773.1 |
More Information
Set Membership: |
|

Genomic Map