Assay Details
Target Gene Details
Entrez Gene ID: | 77480 |
Gene Name: | kinase D-interacting substrate 220 |
Gene Aliases: |
3110039L19Rik, AI194387, AI316525, C330002I19Rik, mKIAA1250 |
Location: |
Chr.12:24974876-25063152 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kidins220 | NM_001081378.1 | 24 | 3427 | NP_001074847.1 |
| AK039537.1 | 3 | 319 | ||
| AK083260.1 | 18 | 2751 | BAE43383.1 | |
| AK122478.1 | 21 | 3018 | BAC65760.1 | |
| BC044772.2 | 19 | 2783 | AAH44772.1 | |
| BC046467.1 | 19 | 2783 | AAH46467.1 | |
| BC094243.1 | 2 | 160 | AAH94243.1 | |
| BC157898.1 | 24 | 3311 | ||
| KJ812113.1 | 1 | 71 | ||
| KJ812114.1 | 1 | 71 | ||
| KJ812115.1 | 1 | 71 | ||
| KJ812116.1 | 1 | 71 | ||
| KJ812117.1 | 1 | 71 | ||
| KJ812118.1 | 1 | 71 |

Genomic Map