Assay Details
Target Gene Details
Entrez Gene ID: | 71693 |
Gene Name: | collectin sub-family member 11 |
Gene Aliases: |
1010001H16Rik |
Location: |
Chr.12:28594172-28623454 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Colec11 | NM_027866.1 | NP_082142.1 | ||
| AK003121.1 | BAB22581.1 | |||
| BC104380.1 | ||||
| BC104381.1 |
More Information
Set Membership: |
|

Genomic Map