Assay Details
Target Gene Details
Entrez Gene ID: | 217674 |
Gene Name: | glycoprotein hormone beta 5 |
Gene Aliases: |
OGH, Zlut1 |
Location: |
Chr.12:75411720-75416781 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gphb5 | NM_175644.3 | 2 | 217 | NP_783575.2 |
| AF467771.1 | 1 | 103 | AAO33391.1 | |
| AK078727.1 | 2 | 218 | ||
| BC111115.1 | 1 | 58 | AAI11116.1 | |
| BC139462.1 | 2 | 157 | ||
| BC139463.1 | 2 | 157 |
More Information
Set Membership: |
|

Genomic Map