Assay Details
Target Gene Details
Entrez Gene ID: | 17286 |
Gene Name: | mesenchyme homeobox 2 |
Gene Aliases: |
AI528662, Gax, Mox-2, Mox2 |
Location: |
Chr.12:37108546-37179528 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Meox2 | NM_008584.3 | NP_032610.1 | ||
| AK028352.1 | BAC25898.1 | |||
| AK031152.1 | BAC27282.1 | |||
| AK142690.1 | BAE25163.1 | |||
| BC002076.1 | AAH02076.1 | |||
| Z16406.1 | CAA78899.1 |
More Information
Set Membership: |
|

Genomic Map