Assay Details
Target Gene Details
Entrez Gene ID: | 217707 |
Gene Name: | coenzyme Q6 homolog (yeast) |
Gene Aliases: |
5930427M12Rik, AW742344 |
Location: |
Chr.12:84361756-84373796 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Coq6 | NM_172582.3 | 1 | 166 | NP_766170.2 |
| AK077863.1 | 1 | 90 | BAC37039.1 | |
| AK131889.1 | 1 | 128 | BAE20853.1 | |
| AK167380.1 | 1 | 94 | BAE39473.1 | |
| AK170311.1 | 1 | 94 | ||
| BC024135.1 | 1 | 67 | AAH24135.1 | |
| BC051055.1 | 1 | 82 | AAH51055.1 | |
| BC132168.1 | 1 | 166 | ||
| BC132170.1 | 1 | 166 |
More Information
Set Membership: |
|

Genomic Map