Assay Details
Target Gene Details
Entrez Gene ID: | 238333 |
Gene Name: | sterile alpha motif domain containing 15 |
Gene Aliases: |
Gm263 |
Location: |
Chr.12:87196168-87213541 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Samd15 | NM_001290288.1 | 3 | 1755 | NP_001277217.1 |
| CJ060055.1 | 3 | 336 |

Genomic Map