Assay Details
Target Gene Details
Entrez Gene ID: | 18217 |
Gene Name: | neurotensin receptor 2 |
Gene Aliases: |
NT2R, NTRL |
Location: |
Chr.12:16653452-16660236 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntsr2 | NM_008747.2 | NP_032773.2 | ||
| AB041826.1 | BAB79256.1 | |||
| AB041827.1 | BAB79257.1 | |||
| AK005368.1 | ||||
| AK081073.1 | ||||
| AK158943.1 | BAE34736.1 | |||
| BC141019.1 | ||||
| U51908.1 | AAB17285.1 |
More Information
Set Membership: |
|

Genomic Map