Assay Details
Target Gene Details
Entrez Gene ID: | 17420 |
Gene Name: | menage a trois 1 |
Gene Aliases: |
E130115E11Rik, MAT1, P36 |
Location: |
Chr.12:73112184-73273988 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mnat1 | NM_008612.2 | NP_032638.2 | ||
| AK008134.1 | BAB25483.1 | |||
| AK008249.1 | BAB25556.1 | |||
| AK011833.1 | ||||
| AK029174.1 | ||||
| AK048499.1 | 2 | 323 | ||
| AK155859.1 | BAE33464.1 | |||
| AK157721.1 | BAE34168.1 | |||
| BC089023.1 | ||||
| BC115630.2 | ||||
| BC115631.1 | ||||
| U35249.1 | AAA91741.1 |
More Information
Set Membership: |
|

Genomic Map