Assay Details
Target Gene Details
Entrez Gene ID: | 100040724 |
Gene Name: | miRNA containing gene |
Gene Aliases: |
2810474H01Rik, AI852992, Gm2928, Meg9, mir-410 |
Location: |
Chr.12:109734981-109749458 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mirg | NR_028265.1 | |||
| AF498297.1 | ||||
| AF498298.1 | ||||
| AJ517767.1 | ||||
| AK013406.1 | ||||
| AK081334.1 | 1 | 383 | ||
| BF465610.1 |

Genomic Map