Assay Details
Target Gene Details
Entrez Gene ID: | 52668 |
Gene Name: | interferon, alpha-inducible protein 27 |
Gene Aliases: |
1110013J02Rik, 2900026P10Rik, D12Ertd647e, ISG12a, Ifi27l1 |
Location: |
Chr.12:103434189-103440245 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi27 | NM_026790.2 | NP_081066.2 | ||
| NM_194066.2 | NP_919042.2 | |||
| NM_194067.2 | NP_919043.1 | |||
| NM_194068.2 | NP_919044.1 | |||
| NM_194069.2 | NP_919045.1 | |||
| AJ566111.1 | CAD92753.1 | |||
| AJ566112.1 | CAD92754.1 | |||
| AJ566113.1 | CAD92755.1 | |||
| AJ566114.1 | CAD92756.1 | |||
| AJ566115.1 | CAD92757.1 | |||
| AK003665.1 | BAB22924.1 | |||
| AK075797.2 | BAC35966.2 | |||
| AK146231.1 | BAE26997.1 | |||
| AK169926.1 | BAE41464.1 | |||
| AK187394.1 | ||||
| BC125301.1 | ||||
| BC125636.1 | ||||
| BC128276.1 | ||||
| BY130468.1 | ||||
| BY130471.1 |
Target Gene Details
Entrez Gene ID: | 76933 |
Gene Name: | interferon, alpha-inducible protein 27 like 2A |
Gene Aliases: |
2310061N23Rik, Ifi27, Isg12, Isg12(b1) |
Location: |
Chr.12:103433873-103443680 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ifi27l2a | NM_001281830.1 | NP_001268759.1 | ||
| AK040279.1 |
More Information
Set Membership: |
|

Genomic Map