Assay Details
Target Gene Details
Entrez Gene ID: | 12313 |
Gene Name: | calmodulin 1 |
Gene Aliases: |
AI256814, AI327027, AI461935, AL024000, CaM, Calm |
Location: |
Chr.12:100199541-100209806 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
More Information
Set Membership: |
|

Genomic Map