Assay Details
Target Gene Details
Entrez Gene ID: | 217869 |
Gene Name: | eukaryotic translation initiation factor 5 |
Gene Aliases: |
2810011H21Rik, D12Ertd549e |
Location: |
Chr.12:111538101-111546753 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5 | NM_173363.5 | NP_775539.1 | ||
| NM_178041.2 | NP_829887.1 | |||
| AI391022.1 |
More Information
Set Membership: |
|

Genomic Map