Assay Details
Target Gene Details
Entrez Gene ID: | 68149 |
Gene Name: | OTU domain, ubiquitin aldehyde binding 2 |
Gene Aliases: |
2010015L18Rik, 4930586I02Rik, AI413508, AW557219, OTB2, OTU2 |
Location: |
Chr.12:103376894-103406351 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otub2 | NM_001177841.1 | 4 | 777 | NP_001171312.1 |
| NM_026580.4 | 3 | 351 | NP_080856.1 | |
| NR_033587.1 | 3 | 633 | ||
| AK006054.1 | 3 | 273 | BAB24385.1 | |
| AK006346.1 | 3 | 303 | BAB24539.1 | |
| AK006469.1 | 3 | 300 | BAB24603.1 | |
| AK008258.2 | 3 | 235 | ||
| AK016449.1 | 3 | 335 | ||
| AK019830.1 | 3 | 328 | BAB31872.1 | |
| AK076336.1 | 4 | 780 | ||
| AK158758.1 | 3 | 634 | BAE34645.1 | |
| AK160505.1 | 3 | 330 | BAE35829.1 | |
| BC049551.1 | 3 | 334 | AAH49551.1 | |
| BY283897.1 | 3 | 351 |
More Information
Set Membership: |
|

Genomic Map