Assay Details
Target Gene Details
Entrez Gene ID: | 217869 |
Gene Name: | eukaryotic translation initiation factor 5 |
Gene Aliases: |
2810011H21Rik, D12Ertd549e |
Location: |
Chr.12:111538101-111546753 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif5 | NM_173363.5 | NP_775539.1 | ||
| NM_178041.2 | NP_829887.1 | |||
| AK076403.1 | BAC36325.1 | |||
| AK159232.1 | BAE34918.1 | |||
| AK160828.1 | ||||
| AK163368.1 | BAE37319.1 | |||
| AK166775.1 | BAE39011.1 | |||
| AK167331.1 | BAE39433.1 | |||
| AK172048.1 | BAE42798.1 | |||
| BC023074.1 | ||||
| BC025557.1 | ||||
| BC039275.1 | AAH39275.1 |
More Information
Set Membership: |
|

Genomic Map