Assay Details
Target Gene Details
Entrez Gene ID: | 110616 |
Gene Name: | ataxin 3 |
Gene Aliases: |
2210008M02Rik, AI463012, AI647473, ATX3, MJD1, Mjd, Sca3, ataxin-3 |
Location: |
Chr.12:101918901-101958800 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atxn3 | NM_001167914.1 | NP_001161386.1 | ||
| NM_029705.3 | NP_083981.2 | |||
| AF537188.1 | AAQ10750.1 | |||
| AK008675.1 | BAB25825.3 | |||
| AK030842.1 | BAC27155.1 | |||
| BC049743.1 | AAH49743.1 | |||
| BC087880.1 | AAH87880.1 |
More Information
Set Membership: |
|

Genomic Map