Assay Details
Target Gene Details
Entrez Gene ID: | 68339 |
Gene Name: | coiled-coil domain containing 88C |
Gene Aliases: |
0610010D24Rik, AV000935, AW324073, Daple, mKIAA1509 |
Location: |
Chr.12:100911521-101028983 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc88c | NM_026681.4 | NP_080957.2 | ||
| AK173188.1 | BAD32466.1 | |||
| AY339881.1 | AAR08446.1 | |||
| BC043115.1 | AAH43115.1 | |||
| BC063255.1 | AAH63255.1 |
More Information
Set Membership: |
|

Genomic Map