Assay Details
Target Gene Details
Entrez Gene ID: | 217615 |
Gene Name: | CTAGE family, member 5 |
Gene Aliases: |
D12Bwg0579e, Mea6, Mgea, Mgea6 |
Location: |
Chr.12:59129740-59190220 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ctage5 | NM_001165253.1 | NP_001158725.1 | ||
| NM_001165254.1 | NP_001158726.1 | |||
| NM_146034.3 | NP_666146.3 | |||
| AK029466.1 | ||||
| AK053010.1 | ||||
| AK080312.1 | ||||
| AK087188.1 | ||||
| AK157358.1 | BAE34063.1 | |||
| AK186274.1 | ||||
| AK188462.1 | ||||
| AK208297.1 | ||||
| BC026864.1 | AAH26864.1 | |||
| BQ571563.1 |
More Information
Set Membership: |
|

Genomic Map