Assay Details
Target Gene Details
Entrez Gene ID: | 328108 |
Gene Name: | family with sequence similarity 179, member B |
Gene Aliases: |
A430041B07Rik, BC062107, C130034K06, mKIAA0423 |
Location: |
Chr.12:64965742-65022573 on Build GRCm38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam179b | NM_177805.3 | NP_808473.2 | ||
| AK084470.1 | 4 | 508 | ||
| AK172948.1 | BAD32226.1 | |||
| BC157958.1 | ||||
| BC158008.1 | 8 | 3348 | ||
| BC172117.1 |
More Information
Set Membership: |
|

Genomic Map