Assay Details
Target Gene Details
Entrez Gene ID: | 13382 |
Gene Name: | dihydrolipoamide dehydrogenase |
Gene Aliases: |
- |
Location: |
Chr.12:31331562-31351471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dld | NM_007861.5 | NP_031887.2 | ||
| AK117104.1 | BAE43405.1 | |||
| AK135807.1 | ||||
| AK136193.1 | BAE22867.1 | |||
| AK153399.1 | BAE31961.1 | |||
| AK167885.1 | BAE39898.1 | |||
| AK168157.1 | ||||
| AK168875.1 | BAE40693.1 | |||
| BC003368.1 | AAH03368.1 | |||
| BY251665.1 | ||||
| BY282701.1 | ||||
| U73445.1 | AAC53170.1 |
More Information
Set Membership: |
|

Genomic Map