Assay Details
Target Gene Details
Entrez Gene ID: | 13386 |
Gene Name: | delta-like 1 homolog (Drosophila) |
Gene Aliases: |
AW742678, DLK-1, DlkI, FA1, Ly107, Peg9, SCP1, ZOG, pG2, pref-1 |
Location: |
Chr.12:109452823-109463336 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlk1 | NR_033813.1 | 1 | 346 | |
| CV562715.1 | 1 | 320 |

Genomic Map