Assay Details
Target Gene Details
Entrez Gene ID: | 70858 |
Gene Name: | RIKEN cDNA 4921511I17 gene |
Gene Aliases: |
- |
Location: |
Chr.12:12351522-12392475 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4921511I17Rik | NR_045829.1 | |||
| AK014875.1 |
Target Gene Details
Entrez Gene ID: | 76820 |
Gene Name: | family with sequence similarity 49, member A |
Gene Aliases: |
2410157M17Rik, 9630047E15, D12Ertd553e |
Location: |
Chr.12:12262129-12379058 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam49a | NM_001146119.1 | NP_001139591.1 | ||
| NM_029758.4 | NP_084034.2 | |||
| AK010821.1 | BAB27204.2 | |||
| AK047142.1 | BAC32969.1 | |||
| BC049664.1 | AAH49664.1 | |||
| BC052465.1 | AAH52465.1 |
More Information
Set Membership: |
|

Genomic Map