Assay Details
Target Gene Details
Entrez Gene ID: | 16777 |
Gene Name: | laminin B1 |
Gene Aliases: |
C77966, C80098, C81607, D130003D08Rik, Lamb-1, Lamb1-1 |
Location: |
Chr.12:31265148-31329644 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamb1 | NM_008482.2 | NP_032508.2 | ||
| AK143079.1 | ||||
| AK147313.1 | BAE27840.1 | |||
| BC121790.1 | ||||
| BC150809.1 | ||||
| M15525.1 | AAA39407.1 |
More Information
Set Membership: |
|

Genomic Map