Assay Details
Target Gene Details
Entrez Gene ID: | 11881 |
Gene Name: | arylsulfatase B |
Gene Aliases: |
1110007C02Rik, AI480648, As-1, As-1r, As-1s, As-1t, As1, As1-r, As1-s, As1-t, Asr-1, Ast-1 |
Location: |
Chr.13:93771023-93943016 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arsb | NM_009712.3 | NP_033842.3 | ||
| AK140163.1 | ||||
| AK154098.1 | BAE32375.1 | |||
| AK158312.1 | BAE34455.1 | |||
| AK189106.1 | ||||
| AK190270.1 | ||||
| AK191477.1 | ||||
| AK195452.1 | ||||
| AK198200.1 | ||||
| AK205135.1 | ||||
| AK211718.1 | ||||
| AK212274.1 | ||||
| AK212990.1 |

Genomic Map