Assay Details
Target Gene Details
Entrez Gene ID: | 66109 |
Gene Name: | tetraspanin 13 |
Gene Aliases: |
1100001I23Rik, NET-6, Tm4sf13 |
Location: |
Chr.12:36014555-36042478 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tspan13 | NM_025359.3 | NP_079635.1 | ||
| AK008175.1 | BAB25510.1 | |||
| AK012571.1 | ||||
| AK090217.1 | BAC41139.1 | |||
| AK090322.1 | BAC41170.1 | |||
| AK158761.1 | BAE34648.1 | |||
| AK165768.1 | BAE38371.1 | |||
| AK167495.1 | BAE39572.1 | |||
| AK167979.1 | BAE39971.1 | |||
| BC018317.1 | AAH18317.1 |

Genomic Map