Assay Details
Target Gene Details
Entrez Gene ID: | 64075 |
Gene Name: | SPARC related modular calcium binding 1 |
Gene Aliases: |
2600002F22Rik, AI848508, SRG |
Location: |
Chr.12:81026808-81186414 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smoc1 | NM_001146217.1 | NP_001139689.1 | ||
| NM_022316.2 | NP_071711.2 | |||
| AF070470.1 | AAD41590.1 | |||
| AK040931.1 | BAC30750.1 | |||
| BC031804.1 | AAH31804.1 |
More Information
Set Membership: |
|

Genomic Map