Assay Details
Target Gene Details
Entrez Gene ID: | 268534 |
Gene Name: | syntrophin, gamma 2 |
Gene Aliases: |
2210008K22Rik, 9530013L23, BB121248 |
Location: |
Chr.12:30174480-30373392 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntg2 | NM_172951.3 | 10 | 932 | NP_766539.2 |
| AF367760.1 | 10 | 787 | AAK53400.1 | |
| AK008673.1 | 10 | 840 | ||
| AK035310.1 | 10 | 886 | BAC29026.1 | |
| AK087560.1 | 10 | 882 | ||
| AK137345.1 | 10 | 891 | ||
| BC138956.1 | 10 | 846 |

Genomic Map