Assay Details
Target Gene Details
Entrez Gene ID: | 66132 |
Gene Name: | RIKEN cDNA 1110008L16 gene |
Gene Aliases: |
Kiaa0391, Mrpp3, mKIAA0391 |
Location: |
Chr.12:55302637-55382491 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1110008L16Rik | NM_025373.1 | 1 | 328 | NP_079649.1 |
| AK045209.1 | 1 | 216 | ||
| AK151976.1 | 1 | 328 | BAE30844.1 | |
| AK152028.1 | 1 | 328 | BAE30887.1 |
Target Gene Details
Entrez Gene ID: | 59032 |
Gene Name: | protein phosphatase 2, regulatory subunit B'', gamma |
Gene Aliases: |
4930511A21Rik, 5730412A08Rik, G4-1, G5pr |
Location: |
Chr.12:55280809-55303005 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r3c | NM_021529.3 | NP_067504.2 | ||
| AB041577.1 | BAA95061.1 | |||
| AK028063.1 | ||||
| AK033405.1 | ||||
| AK168298.1 |
More Information
Set Membership: |
|

Genomic Map