Assay Details
Target Gene Details
Entrez Gene ID: | 59032 |
Gene Name: | protein phosphatase 2, regulatory subunit B'', gamma |
Gene Aliases: |
4930511A21Rik, 5730412A08Rik, G4-1, G5pr |
Location: |
Chr.12:55280809-55303005 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp2r3c | NM_021529.3 | 3 | 353 | NP_067504.2 |
| AB041577.1 | 3 | 342 | BAA95061.1 | |
| AJ238247.2 | 2 | 180 | CAB88038.2 | |
| AK013403.1 | 2 | 149 | BAB28835.1 | |
| AK015746.1 | 2 | 181 | ||
| AK017550.1 | 2 | 136 | ||
| AK028063.1 | 3 | 318 | ||
| AK028254.1 | 2 | 272 | BAC25845.1 | |
| AK033405.1 | 3 | 351 | ||
| AK040589.1 | 2 | 181 | BAC30637.1 | |
| AK087572.1 | 2 | 176 | BAC39933.1 | |
| AK088240.1 | 2 | 262 | BAC40229.1 | |
| AK168298.1 | 3 | 354 | ||
| BC138295.1 | 2 | 149 | ||
| BC138296.1 | 2 | 149 |
More Information
Set Membership: |
|

Genomic Map