Assay Details
Target Gene Details
Entrez Gene ID: | 268534 |
Gene Name: | syntrophin, gamma 2 |
Gene Aliases: |
2210008K22Rik, 9530013L23, BB121248 |
Location: |
Chr.12:30174480-30373392 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sntg2 | NM_172951.3 | NP_766539.2 | ||
| AF367760.1 | AAK53400.1 | |||
| AK008673.1 | ||||
| AK035310.1 | BAC29026.1 | |||
| AK087560.1 | ||||
| AK137345.1 | ||||
| BC138956.1 |

Genomic Map