Assay Details
Target Gene Details
Entrez Gene ID: | 21809 |
Gene Name: | transforming growth factor, beta 3 |
Gene Aliases: |
Tgfb-3 |
Location: |
Chr.12:86056743-86079041 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tgfb3 | NM_009368.3 | NP_033394.2 | ||
| AK132519.1 | BAE21215.1 | |||
| AK145304.1 | BAE26357.1 | |||
| AK148385.1 | BAE28523.1 | |||
| AK161328.1 | BAE36326.1 | |||
| AK162560.1 | BAE36968.1 | |||
| BC005513.1 | AAH05513.1 | |||
| BC014690.1 | AAH14690.1 | |||
| BC108426.1 | AAI08427.1 | |||
| M32745.1 | AAA40422.1 |
More Information
Set Membership: |
|

Genomic Map