Assay Details
Target Gene Details
Entrez Gene ID: | 22375 |
Gene Name: | tryptophanyl-tRNA synthetase |
Gene Aliases: |
TrpRS, WRS |
Location: |
Chr.12:108860030-108894174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wars | NM_001164314.1 | NP_001157786.1 | ||
| NM_001164488.1 | NP_001157960.1 | |||
| NM_011710.3 | NP_035840.3 | |||
| AK004541.1 | BAB23357.1 | |||
| AK007754.1 | ||||
| AK150090.1 | BAE29299.1 | |||
| AK150267.1 | ||||
| AK152966.1 | BAE31627.1 | |||
| AK153337.1 | ||||
| AK179345.1 | ||||
| AK179848.1 | ||||
| AK183767.1 | ||||
| AK185391.1 | ||||
| AK190023.1 | ||||
| AK190625.1 | ||||
| AK192489.1 | ||||
| AK193136.1 | ||||
| AK196899.1 | ||||
| AK200381.1 | ||||
| AK200730.1 | ||||
| AK201963.1 | ||||
| AK203296.1 | ||||
| BC003450.1 | AAH03450.1 | |||
| BC046232.1 | AAH46232.1 | |||
| CF731228.1 | ||||
| CT010258.1 | CAJ18466.1 |
More Information
Set Membership: |
|

Genomic Map