Assay Details
Target Gene Details
Entrez Gene ID: | 22375 |
Gene Name: | tryptophanyl-tRNA synthetase |
Gene Aliases: |
TrpRS, WRS |
Location: |
Chr.12:108860030-108894174 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wars | NM_001164314.1 | 7 | 957 | NP_001157786.1 |
| NM_001164488.1 | 7 | 890 | NP_001157960.1 | |
| NM_011710.3 | 7 | 903 | NP_035840.3 | |
| AK004541.1 | 7 | 891 | BAB23357.1 | |
| AK007754.1 | 5 | 519 | ||
| AK150090.1 | 8 | 921 | BAE29299.1 | |
| AK150267.1 | 8 | 1005 | ||
| AK152966.1 | 7 | 921 | BAE31627.1 | |
| AK153337.1 | 7 | 928 | ||
| AK179345.1 | ||||
| BC003450.1 | 7 | 889 | AAH03450.1 | |
| BC046232.1 | 7 | 896 | AAH46232.1 | |
| CF731228.1 | 4 | 409 | ||
| CT010258.1 | 6 | 789 | CAJ18466.1 |
More Information
Set Membership: |
|

Genomic Map