Assay Details
Target Gene Details
Entrez Gene ID: | 52708 |
Gene Name: | zinc finger protein 410 |
Gene Aliases: |
Apa1, D12Ertd748e, Znf410 |
Location: |
Chr.12:84315928-84344439 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp410 | NM_001252582.1 | 7 | 1355 | NP_001239511.1 |
| NM_001252583.1 | 7 | 1320 | NP_001239512.1 | |
| NM_144833.3 | 7 | 1320 | NP_659082.1 | |
| NR_045546.1 | 7 | 1288 | ||
| NR_045615.1 | 5 | 1004 | ||
| AF295806.1 | 7 | 1082 | AAL82191.1 | |
| AK033090.1 | 7 | 1097 | BAC28148.1 | |
| AK051939.1 | 5 | 720 | ||
| AK053001.1 | 7 | 1045 | BAC35235.1 | |
| AK083003.1 | 7 | 1063 | ||
| AK132345.1 | 7 | 1058 | ||
| AK139797.1 | 6 | 874 | ||
| BC021528.1 | 7 | 1075 | AAH21528.1 | |
| BC052080.1 | 7 | 1052 | AAH52080.1 |
More Information
Set Membership: |
|

Genomic Map