Assay Details
Target Gene Details
Entrez Gene ID: | 271144 |
Gene Name: | ankyrin repeat and death domain containing 1B |
Gene Aliases: |
9330128J19Rik |
Location: |
Chr.13:96415030-96471354 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankdd1b | NM_001042714.1 | 1 | 78 | NP_001036179.1 |
| BC112417.1 | 1 | 78 |

Genomic Map