Assay Details
Target Gene Details
Entrez Gene ID: | 11992 |
Gene Name: | AU RNA binding protein/enoyl-coenzyme A hydratase |
Gene Aliases: |
C77140, W91705 |
Location: |
Chr.13:52835102-52929706 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Auh | NM_016709.2 | 3 | 342 | NP_057918.2 |
| AK003219.1 | 3 | 321 | ||
| AK003929.1 | 3 | 322 | BAB23078.1 | |
| AK019978.1 | 3 | 319 | BAB31947.1 | |
| AK134886.1 | 3 | 314 | ||
| AK147855.1 | 3 | 346 | ||
| AK169661.1 | 3 | 320 | BAE41283.1 | |
| BC026525.1 | 3 | 280 | AAH26525.1 | |
| BC049597.1 | 3 | 326 | AAH49597.1 | |
| BY741014.1 | 3 | 343 |
More Information
Set Membership: |
|

Genomic Map