Assay Details
Target Gene Details
Entrez Gene ID: | 11992 |
Gene Name: | AU RNA binding protein/enoyl-coenzyme A hydratase |
Gene Aliases: |
C77140, W91705 |
Location: |
Chr.13:52835102-52929706 on Build GRCm38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Auh | NM_016709.2 | NP_057918.2 | ||
| AK003219.1 | ||||
| BC026525.1 | AAH26525.1 | |||
| BC049597.1 | AAH49597.1 |
More Information
Set Membership: |
|

Genomic Map