Assay Details
Target Gene Details
Entrez Gene ID: | 66354 |
Gene Name: | SNW domain containing 1 |
Gene Aliases: |
2310008B08Rik, AW048543, NCoA-62, SKIP, Skiip |
Location: |
Chr.12:87449910-87472299 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snw1 | NM_025507.2 | NP_079783.2 | ||
| AK009218.1 | BAB26144.1 | |||
| AK012384.1 | BAB28203.2 | |||
| AK166163.1 | BAE38605.1 | |||
| AK167695.1 | ||||
| AX775977.1 | CAE11688.1 | |||
| AX775979.1 | CAE11689.1 |
More Information
Set Membership: |
|

Genomic Map