Assay Details
Target Gene Details
Entrez Gene ID: | 68581 |
Gene Name: | transmembrane emp24-like trafficking protein 10 (yeast) |
Gene Aliases: |
1110014C03Rik, Tmp21, p23, p24delta1 |
Location: |
Chr.12:85340614-85374717 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmed10 | NM_026775.4 | NP_081051.1 | ||
| AK003676.1 | BAB22932.1 | |||
| AK084117.1 | BAC39119.1 | |||
| AK150605.1 | BAE29697.1 | |||
| AK152643.1 | BAE31382.1 | |||
| AK152656.1 | BAE31393.1 | |||
| AK186798.1 | ||||
| AK191153.1 | ||||
| AK198152.1 | ||||
| AK201939.1 | ||||
| AK206502.1 | ||||
| AK207697.1 | ||||
| AK216393.1 | ||||
| BC006774.1 | AAH06774.1 | |||
| BC064755.1 | AAH64755.1 | |||
| BC085097.1 | AAH85097.1 |
More Information
Set Membership: |
|

Genomic Map