Assay Details
Target Gene Details
Entrez Gene ID: | 217864 |
Gene Name: | REST corepressor 1 |
Gene Aliases: |
5730409O11, 6720480E22Rik, AU042633, D12Wsu95e, Rocr1, mKIAA0071 |
Location: |
Chr.12:111038236-111115901 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcor1 | NM_198023.2 | 8 | 693 | NP_932140.1 |
| AB093210.1 | 4 | 394 | BAC41394.1 | |
| AK077445.1 | 2 | 134 | BAC36804.1 | |
| BC042731.1 | 8 | 693 | AAH42731.1 |

Genomic Map