Assay Details
Target Gene Details
Entrez Gene ID: | 77318 |
Gene Name: | ankyrin repeat domain 55 |
Gene Aliases: |
C030011J08Rik |
Location: |
Chr.13:112288451-112384002 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd55 | NM_001168403.1 | 3 | 694 | NP_001161875.1 |
| NM_001168404.1 | 3 | 231 | NP_001161876.1 | |
| NM_001168405.1 | 2 | 588 | NP_001161877.1 | |
| NM_029898.3 | 2 | 588 | NP_084174.2 | |
| AK039553.1 | 2 | 589 | BAC30379.1 | |
| AK045480.1 | 2 | 222 | BAC32388.1 | |
| AK081197.1 | 3 | 232 | BAC38161.1 | |
| AK138273.1 | 2 | 243 | BAE23605.1 | |
| AK140417.1 | 3 | 557 | ||
| AK155092.1 | ||||
| AK171119.1 | BAE42261.1 | |||
| BC152317.1 | 2 | 215 | ||
| BC152536.1 | 2 | 215 |
More Information
Set Membership: |
|

Genomic Map