Assay Details
Target Gene Details
Entrez Gene ID: | 77318 |
Gene Name: | ankyrin repeat domain 55 |
Gene Aliases: |
C030011J08Rik |
Location: |
Chr.13:112288451-112384002 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd55 | NM_001168403.1 | NP_001161875.1 | ||
| NM_001168404.1 | NP_001161876.1 | |||
| NM_001168405.1 | NP_001161877.1 | |||
| NM_029898.3 | NP_084174.2 | |||
| AK039553.1 | BAC30379.1 | |||
| AK045480.1 | BAC32388.1 | |||
| AK081197.1 | BAC38161.1 | |||
| AK138273.1 | BAE23605.1 | |||
| AK140417.1 | ||||
| AK155092.1 | ||||
| AK171119.1 | BAE42261.1 | |||
| BC152317.1 | ||||
| BC152536.1 |
More Information
Set Membership: |
|

Genomic Map